Learn More
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 368320250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(Trifluoromethyl)trimethylsilane | |
81290-20-2,109-99-9 | |
8.74,92.85 | |
0.8950g/mL | |
-20°C | |
C5H11F3Si | |
MFCD00145454 | |
0.895 | |
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent | |
CC(C)(C)[SiH2]C(F)(F)F | |
156.22 | |
142.2 |
0.5M solution in THF | |
7.15,91.26 | |
Colorless | |
40.0°C | |
Glass bottle | |
(CH3)3SiCF3 | |
25 mL | |
15, 9851 | |
MTYSDPUZDZURPP-UHFFFAOYSA-N | |
trimethyl(trifluoromethyl)silane | |
552549 | |
Liquid |
Chemical Identifiers
81290-20-2 | |
156.22 | |
MTYSDPUZDZURPP-UHFFFAOYSA-N | |
552549 |
C5H11F3Si | |
MFCD00145454 | |
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent | |
CC(C)(C)[SiH2]C(F)(F)F |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only