missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triglycidyl Isocyanurate 98.0+%, TCI America™
Supplier: TCI America I0428100G
Specifications
| Triglycidyl Isocyanurate | |
| 108°C | |
| C12H15N3O6 | |
| 100 g | |
| Isocyanuric Acid Tris(2,3-epoxypropyl) Ester, Isocyanuric Acid Triglycidyl Ester, Tris(2,3-epoxypropyl) Isocyanurate | |
| C1C(O1)CN2C(=O)N(C(=O)N(C2=O)CC3CO3)CC4CO4 | |
| 297.267 | |
| 297.27 | |
| Crystalline Powder |
| 2451-62-9 | |
| White | |
| MFCD00080670 | |
| 2811 | |
| OUPZKGBUJRBPGC-UHFFFAOYSA-N | |
| 1,3,5-tris(oxiran-2-ylmethyl)-1,3,5-triazinane-2,4,6-trione | |
| 17142 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 2451-62-9 | |
| 297.267 | |
| OUPZKGBUJRBPGC-UHFFFAOYSA-N | |
| 17142 | |
| C1C(O1)CN2C(=O)N(C(=O)N(C2=O)CC3CO3)CC4CO4 |
| C12H15N3O6 | |
| MFCD00080670 | |
| Isocyanuric Acid Tris(2,3-epoxypropyl) Ester, Isocyanuric Acid Triglycidyl Ester, Tris(2,3-epoxypropyl) Isocyanurate | |
| 1,3,5-tris(oxiran-2-ylmethyl)-1,3,5-triazinane-2,4,6-trione |
Safety and Handling
EINECSNumber : (5)-1052
RTECSNumber : XZ1994900
TSCA : Yes