Learn More
Triisopropanolamine, 98%, Thermo Scientific Chemicals
$62.41 - $114.39
Chemical Identifiers
CAS | 122-20-3 |
---|---|
Molecular Formula | C9H21NO3 |
Molecular Weight (g/mol) | 191.27 |
MDL Number | MFCD00004533 |
InChI Key | SLINHMUFWFWBMU-UHFFFAOYSA-N |
Synonym | triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891 |
PubChem CID | 24730 |
IUPAC Name | 1-[bis(2-hydroxypropyl)amino]propan-2-ol |
SMILES | CC(CN(CC(C)O)CC(C)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC245761000
|
Thermo Scientific Chemicals
245761000 |
100 g | Glass bottle |
Each for $62.41
|
|
||||
AC245760010
|
Thermo Scientific Chemicals
245760010 |
1 kg | Glass bottle |
Each for $114.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
122-20-3 | |
191.27 | |
SLINHMUFWFWBMU-UHFFFAOYSA-N | |
24730 | |
CC(CN(CC(C)O)CC(C)O)O |
C9H21NO3 | |
MFCD00004533 | |
triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891 | |
1-[bis(2-hydroxypropyl)amino]propan-2-ol |
Specifications
122-20-3 | |
White to Yellow | |
160°C | |
98% | |
C9H21NO3 | |
MFCD00004533 | |
04,762 | |
triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891 | |
SLINHMUFWFWBMU-UHFFFAOYSA-N | |
1-[bis(2-hydroxypropyl)amino]propan-2-ol | |
24730 | |
191.27 | |
Crystalline Low Melting Solid |
48.0°C to 52.0°C | |
190.0°C (23.0 mmHg) | |
Authentic | |
Glass bottle | |
N(CH2CH(OH)CH3)3 | |
100 g | |
17,371; 12,445; 13,46 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether | |
CC(CN(CC(C)O)CC(C)O)O | |
191.27 | |
138 mPa.s (60°C) | |
98% | |
Triisopropanolamine |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Warning
EINECSNumber : 204-528-4
RUO – Research Use Only