missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triisopropylphosphine, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03357209

 View more versions of this product

Catalog No. AA3357209


Explore available promotions

Description

Description

Triisopropylphosphine is used as a ligand in organometallic chemistry. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals. It is also used to produce Chlor(triisopropyl)phosphonium-dimesylamid at temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Triisopropylphosphine
0.839 g/mL
-17°C (1°F)
C9H21P
MFCD00015029
UN2845
triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458
IGNTWNVBGLNYDV-UHFFFAOYSA-N
tri(propan-2-yl)phosphane
80969
≥90%
6476-36-4
176°C to 178°C
Pungent
1.467
10 g
Air Sensitive
Soluble in alkanes.
CC(C)P(C(C)C)C(C)C
160.241
160.24
Liquid

Chemical Identifiers

6476-36-4
160.241
IGNTWNVBGLNYDV-UHFFFAOYSA-N
80969
CC(C)P(C(C)C)C(C)C
C9H21P
MFCD00015029
triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458
tri(propan-2-yl)phosphane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P222-P260-P264b-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H250-H314

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 229-333-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only