missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trilaurin, Thermo Scientific Chemicals

$52.37 - $143.88

Chemical Identifiers

CAS 538-24-9
Molecular Formula C39H74O6
Molecular Weight (g/mol) 639.015
MDL Number MFCD00026559
InChI Key VMPHSYLJUKZBJJ-UHFFFAOYSA-N
Synonym trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
PubChem CID 10851
ChEBI CHEBI:77389
IUPAC Name 2,3-di(dodecanoyloxy)propyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6296206
View Documents
Thermo Scientific Chemicals
J6296206
5 g
Each for $52.37
 
AAJ6296214
View Documents
Thermo Scientific Chemicals
J6296214
25 g
Each for $143.88
 
Description

Description

Trilaurin is used in makeup products, creams and lotions, deodorants, suntan and sunscreen products, hair conditioners, and skin care and skin cleansing products. These are used in cosmetic products as thickening agents and emollients. It is used as a viscosity controlling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trilaurin is used in makeup products, creams and lotions, deodorants, suntan and sunscreen products, hair conditioners, and skin care and skin cleansing products. These are used in cosmetic products as thickening agents and emollients. It is used as a viscosity controlling agent.

Solubility
Soluble in water, ethylacetate- almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

538-24-9
639.015
VMPHSYLJUKZBJJ-UHFFFAOYSA-N
10851
2,3-di(dodecanoyloxy)propyl dodecanoate
C39H74O6
MFCD00026559
trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
CHEBI:77389
CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

Specifications

538-24-9
White
[CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
5 g
trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
VMPHSYLJUKZBJJ-UHFFFAOYSA-N
2,3-di(dodecanoyloxy)propyl dodecanoate
10851
639.01
Trilaurin
47°C
C39H74O6
MFCD00026559
1730452
Soluble in water,ethylacetate- almost transparency.
CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC
639.015
CHEBI:77389
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-687-0

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only