Learn More
Trimellitic anhydride chloride, 98%, Thermo Scientific Chemicals
$70.48 - $170.44
Chemical Identifiers
CAS | 1204-28-0 |
---|---|
Molecular Formula | C9H3ClO4 |
Molecular Weight (g/mol) | 210.569 |
MDL Number | MFCD00005924 |
InChI Key | NJMOHBDCGXJLNJ-UHFFFAOYSA-N |
Synonym | trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride |
PubChem CID | 70998 |
IUPAC Name | 1,3-dioxo-2-benzofuran-5-carbonyl chloride |
SMILES | C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1438214
|
Thermo Scientific Chemicals
L1438214 |
25 g |
Each for $70.48
|
|
|||||
AAL1438222
|
Thermo Scientific Chemicals
L1438222 |
100 g |
Each for $170.44
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1204-28-0 | |
210.569 | |
NJMOHBDCGXJLNJ-UHFFFAOYSA-N | |
70998 | |
C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O |
C9H3ClO4 | |
MFCD00005924 | |
trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride | |
1,3-dioxo-2-benzofuran-5-carbonyl chloride |
Specifications
1204-28-0 | |
C9H3ClO4 | |
25 g | |
610823 | |
trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride | |
C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O | |
210.569 | |
210.57 | |
Trimellitic anhydride chloride |
67°C to 69°C | |
MFCD00005924 | |
UN3261 | |
Moisture sensitive | |
NJMOHBDCGXJLNJ-UHFFFAOYSA-N | |
1,3-dioxo-2-benzofuran-5-carbonyl chloride | |
70998 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 214-874-8
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only