missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 97.0+%, TCI America™
$321.92 - $959.86
Chemical Identifiers
CAS | 85877-79-8 |
---|---|
Molecular Formula | C9H13F9O3Si |
Molecular Weight (g/mol) | 368.271 |
MDL Number | MFCD08275518 |
InChI Key | IJROHELDTBDTPH-UHFFFAOYSA-N |
Synonym | Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane, 1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane |
PubChem CID | 15651659 |
IUPAC Name | trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane |
SMILES | CO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OC)OC |
Chemical Identifiers
85877-79-8 | |
368.271 | |
IJROHELDTBDTPH-UHFFFAOYSA-N | |
15651659 | |
CO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OC)OC |
C9H13F9O3Si | |
MFCD08275518 | |
Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane, 1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane | |
trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane |
Specifications
85877-79-8 | |
43°C | |
MFCD08275518 | |
Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane, 1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane | |
CO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OC)OC | |
368.271 | |
368.27 | |
Liquid |
Colorless | |
C9H13F9O3Si | |
5 g | |
IJROHELDTBDTPH-UHFFFAOYSA-N | |
trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane | |
15651659 | |
≥97.0% (GC) | |
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane |
Safety and Handling
TSCA : No