missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethoxy(methyl)silane 98.0+%, TCI America™

Supplier:  TCI America M0660100ML

 View more versions of this product

Catalog No. M0660100ML


Explore available promotions

Description

Description

Specifications

Specifications

Trimethoxy(methyl)silane
Colorless
C4H12O3Si
100 mL
methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
CO[Si](C)(OC)OC
136.222
136.22
Liquid
1185-55-3
103°C
MFCD00008342
1993
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
trimethoxy(methyl)silane
14456
≥98.0% (GC)

Chemical Identifiers

1185-55-3
136.222
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
14456
CO[Si](C)(OC)OC
C4H12O3Si
MFCD00008342
methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
trimethoxy(methyl)silane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2052&(2)-2053

RTECSNumber : VV4650000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions