missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl 1,3,5-benzenetricarboxylate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2192606

 View more versions of this product

Catalog No. AAB2192606


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trimethyl 1,3,5-benzenetricarboxylate
144°C to 147°C
MFCD00008434
2219698
RGCHNYAILFZUPL-UHFFFAOYSA-N
trimethyl benzene-1,3,5-tricarboxylate
75881
98%
2672-58-4
C12H12O6
5 g
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
252.222
252.22

Chemical Identifiers

2672-58-4
252.222
RGCHNYAILFZUPL-UHFFFAOYSA-N
75881
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
C12H12O6
MFCD00008434
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
trimethyl benzene-1,3,5-tricarboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-215-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only