missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethyl 1,3,5-benzenetricarboxylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2192614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethyl 1,3,5-benzenetricarboxylate | |
144°C to 147°C | |
MFCD00008434 | |
2219698 | |
RGCHNYAILFZUPL-UHFFFAOYSA-N | |
trimethyl benzene-1,3,5-tricarboxylate | |
75881 | |
98% |
2672-58-4 | |
C12H12O6 | |
25 g | |
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766 | |
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC | |
252.222 | |
252.22 |
Chemical Identifiers
2672-58-4 | |
252.222 | |
RGCHNYAILFZUPL-UHFFFAOYSA-N | |
75881 | |
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC |
C12H12O6 | |
MFCD00008434 | |
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766 | |
trimethyl benzene-1,3,5-tricarboxylate |
Safety and Handling
EINECSNumber : 220-215-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only