missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl borate, 99%, Thermo Scientific Chemicals

$39.12 - $89.63

Chemical Identifiers

CAS 121-43-7
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC140121000
SDS View Documents
Thermo Scientific Chemicals
140121000
100 g Glass bottle
Each for $39.12
 
AC140125000
SDS View Documents
Thermo Scientific Chemicals
140125000
500 g Glass bottle
Each for $89.63
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

121-43-7
WRECIMRULFAWHA-UHFFFAOYSA-N
8470
trimethyl borate
MFCD00008346
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
CHEBI:38913
B(OC)(OC)OC

Specifications

121-43-7
Colorless
68.0°C to 69.0°C
Authentic
Glass bottle
1.3562 to 1.3582
MFCD00008346
01, 287
0.915
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethyl borate
8470
CHEBI:38913
99%
Trimethyl borate, 99%
-34.0°C
0.9150g/mL
−8°C
99%
C3H9O3B
(CH3O)3B
100 g
01,673; 13,218
15, 9882
Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids
B(OC)(OC)OC
103.91
0.36 mPa.s (25°C)
103.91
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 204-468-9

RTECSNumber : ED5600000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only