missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl Borate 98.0+%, TCI America™

Supplier:  TCI America B0522500ML

 View more versions of this product

Catalog No. B0522500ML


Explore available promotions

Description

Description

Specifications

Specifications

Trimethyl Borate
-29°C
68°C
MFCD00008346
2416
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethyl borate
8470
103.91
Liquid
121-43-7
Yellow
C3H9BO3
500 mL
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
B(OC)(OC)OC
103.912
CHEBI:38913
≥98.0% (T)

Chemical Identifiers

121-43-7
103.912
WRECIMRULFAWHA-UHFFFAOYSA-N
8470
trimethyl borate
C3H9BO3
MFCD00008346
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
CHEBI:38913
B(OC)(OC)OC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2101&(2)-2114

RTECSNumber : ED5600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions