missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethyl Orthoformate, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor TR124200LTBL
Specifications
149-73-5 | |
Drum | |
200 L | |
Reagent |
1 | |
1.3765-1.3815 | |
0.99 |
Chemical Identifiers
149-73-5 |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.