Learn More
Trimethyl orthoformate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 148661000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Trimethyl orthoformate | |
-53.0°C | |
0.9700g/mL | |
13°C | |
98.5% min. (GC) | |
C4H10O3 | |
CH(OCH3)3 | |
100 g | |
03,313; 04,540; 05,714; 08,507; 10,425; 14,330; 15,342 | |
15, 6980 | |
Solubility in water: 10g/L (hydrolysis). Other solubilities: soluble in organic solvents | |
COC(OC)OC | |
106.12 | |
106.12 | |
Liquid |
149-73-5 | |
Colorless | |
101.0°C to 102.0°C | |
Authentic | |
Glass bottle | |
1.3780 to 1.3800 | |
MFCD00008483 | |
02, 19 | |
0.97 | |
trimethyl orthoformate, methyl orthoformate, methane, trimethoxy, orthoformic acid, trimethyl ester, orthoformic acid methyl ester, orthomravencan methylnaty, methoxymethylal, methylester kyseliny orthomravenci, unii-xam28819yj, orthoformic acid trimethyl ester | |
PYOKUURKVVELLB-UHFFFAOYSA-N | |
trimethoxymethane | |
9005 | |
99% |
Chemical Identifiers
149-73-5 | |
106.12 | |
PYOKUURKVVELLB-UHFFFAOYSA-N | |
9005 | |
COC(OC)OC |
C4H10O3 | |
MFCD00008483 | |
trimethyl orthoformate, methyl orthoformate, methane, trimethoxy, orthoformic acid, trimethyl ester, orthoformic acid methyl ester, orthomravencan methylnaty, methoxymethylal, methylester kyseliny orthomravenci, unii-xam28819yj, orthoformic acid trimethyl ester | |
trimethoxymethane |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 205-745-7
RUO – Research Use Only