missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl orthoformate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 148665000

 View more versions of this product

Catalog No. AC148665000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Trimethyl orthoformate
-53.0°C
0.9700g/mL
13°C
98.5% min. (GC)
C4H10O3
CH(OCH3)3
500 g
03,313; 04,540; 05,714; 08,507; 10,425; 14,330; 15,342
15, 6980
Solubility in water: 10g/L (hydrolysis). Other solubilities: soluble in organic solvents
COC(OC)OC
106.12
106.12
Liquid
149-73-5
Colorless
101.0°C to 102.0°C
Authentic
Glass bottle
1.3780 to 1.3800
MFCD00008483
02, 19
0.97
trimethyl orthoformate, methyl orthoformate, methane, trimethoxy, orthoformic acid, trimethyl ester, orthoformic acid methyl ester, orthomravencan methylnaty, methoxymethylal, methylester kyseliny orthomravenci, unii-xam28819yj, orthoformic acid trimethyl ester
PYOKUURKVVELLB-UHFFFAOYSA-N
trimethoxymethane
9005
99%

Chemical Identifiers

149-73-5
106.12
PYOKUURKVVELLB-UHFFFAOYSA-N
9005
COC(OC)OC
C4H10O3
MFCD00008483
trimethyl orthoformate, methyl orthoformate, methane, trimethoxy, orthoformic acid, trimethyl ester, orthoformic acid methyl ester, orthomravencan methylnaty, methoxymethylal, methylester kyseliny orthomravenci, unii-xam28819yj, orthoformic acid trimethyl ester
trimethoxymethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-745-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only