missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 043282K4

 View more versions of this product

Catalog No. AA43282K4


Explore available promotions

Description

Description

Trimethylamine solution used in research. As flavoring agents and adjuvants. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators, strongly basic anion exchange resins, dye leveling agents and a number of basic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trimethylamine
-2°C
30°C
Unpleasant
MFCD00008327
UN1297
14,9710
Soluble In Water,8.9e+005mg/L.
CN(C)C
59.11
CHEBI:18139
45% w/w aq. soln.
75-50-3
Colorless
−20°C (−4°F)
C3H9N
2.5 L
956566
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
GETQZCLCWQTVFV-UHFFFAOYSA-N
N,N-dimethylmethanamine
1146
59.11
Liquid

Chemical Identifiers

75-50-3
59.11
GETQZCLCWQTVFV-UHFFFAOYSA-N
1146
CN(C)C
C3H9N
MFCD00008327
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
CHEBI:18139
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H314-H302-H332-H335
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H224-H302+H332-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: TRIMETHYLAMINE, AQUEOUS SOLUTIONS

EINECSNumber : 200-875-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only