missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylamine N-Oxide Anhydrous 95.0+%, TCI America™

Supplier:  TCI America T13625G

 View more versions of this product

Catalog No. T13625G


Explore available promotions

Description

Description

Specifications

Specifications

Trimethylamine N-Oxide Anhydrous
222°C
C3H9NO
5 g
UYPYRKYUKCHHIB-UHFFFAOYSA-N
N,N-dimethylmethanamine oxide
1145
75.11
Crystalline Powder
1184-78-7
White-Yellow
MFCD00002048
TMANO
C[N+](C)(C)[O-]
75.111
CHEBI:15724
≥95.0% (T)

Chemical Identifiers

1184-78-7
75.111
UYPYRKYUKCHHIB-UHFFFAOYSA-N
1145
N,N-dimethylmethanamine oxide
C3H9NO
MFCD00002048
TMANO
CHEBI:15724
C[N+](C)(C)[O-]
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions