missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylamine N-Oxide Dihydrate 98.0+%, TCI America™
$93.43 - $858.13
Chemical Identifiers
CAS | 62637-93-8 |
---|---|
Molecular Formula | C3H13NO3 |
Molecular Weight (g/mol) | 111.14 |
MDL Number | MFCD00149077 |
InChI Key | PGFPZGKEDZGJQZ-UHFFFAOYSA-N |
Synonym | trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate |
PubChem CID | 198430 |
IUPAC Name | N,N-dimethylmethanamine oxide dihydrate |
SMILES | O.O.C[N+](C)(C)[O-] |
Chemical Identifiers
62637-93-8 | |
111.14 | |
PGFPZGKEDZGJQZ-UHFFFAOYSA-N | |
198430 | |
O.O.C[N+](C)(C)[O-] |
C3H13NO3 | |
MFCD00149077 | |
trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate | |
N,N-dimethylmethanamine oxide dihydrate |
Specifications
62637-93-8 | |
White | |
MFCD00149077 | |
trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate | |
O.O.C[N+](C)(C)[O-] | |
111.14 | |
75.11 | |
Crystalline Powder |
95°C | |
C3H13NO3 | |
25 g | |
PGFPZGKEDZGJQZ-UHFFFAOYSA-N | |
N,N-dimethylmethanamine oxide dihydrate | |
198430 | |
≥98.0% (T) | |
Trimethylamine N-Oxide Dihydrate |
Safety and Handling
RTECSNumber : YH2850000
TSCA : Yes