missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylhexamethylene Diisocyanate (2,2,4- and 2,4,4- mixture) 97.0+%, TCI America™
$63.38 - $464.14
Chemical Identifiers
| CAS | 28679-16-5 |
|---|---|
| Molecular Formula | C11H18N2O2 |
| MDL Number | MFCD00040422 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T117625ML
|
TCI America
T117625ML |
25 mL |
Each for $63.38
|
|
|||||
|
T1176100ML
|
TCI America
T1176100ML |
100 mL |
Each for $154.71
|
|
|||||
|
T1176500ML
|
TCI America
T1176500ML |
500 mL |
Each for $464.14
|
|
|||||
Chemical Identifiers
| 28679-16-5 | |
| C11H18N2O2 | |
| MFCD00040422 |
Specifications
| 28679-16-5 | |
| C11H18N2O2 | |
| 25 mL | |
| 210.28 | |
| Liquid |
| Yellow | |
| MFCD00040422 | |
| 2328 | |
| ≥97.0% (GC) | |
| Trimethylhexamethylene Diisocyanate (2,2,4- and 2,4,4- mixture) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator