missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylolethane 98.0+%, TCI America™
Supplier: TCI America T047925G
Specifications
Trimethylolethane | |
201°C | |
137°C | |
MFCD00004687 | |
trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane | |
CC(CO)(CO)CO | |
120.148 | |
120.15 | |
Crystalline Powder |
77-85-0 | |
White | |
C5H12O3 | |
25 g | |
QXJQHYBHAIHNGG-UHFFFAOYSA-N | |
2-(hydroxymethyl)-2-methylpropane-1,3-diol | |
6502 | |
≥98.0% (GC) |
Chemical Identifiers
77-85-0 | |
120.148 | |
QXJQHYBHAIHNGG-UHFFFAOYSA-N | |
6502 | |
CC(CO)(CO)CO |
C5H12O3 | |
MFCD00004687 | |
trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane | |
2-(hydroxymethyl)-2-methylpropane-1,3-diol |
Safety and Handling
EINECSNumber : (2)-0243
RTECSNumber : TY7137500
TSCA : Yes