Learn More
Trimethylphosphine, 98%, Thermo Scientific Chemicals
$92.14 - $1285.23
Chemical Identifiers
CAS | 594-09-2 |
---|---|
Molecular Formula | C3H9P |
Molecular Weight (g/mol) | 76.079 |
MDL Number | MFCD00008510 |
InChI Key | YWWDBCBWQNCYNR-UHFFFAOYSA-N |
Synonym | trimethylphosphine, trimethyl phosphine, pme3, trimethylphosphorus, phosphine, trimethyl, me3p, ch3 3p, trimethylphospine, trimethylphosphine?, phosphine,trimethyl |
PubChem CID | 68983 |
ChEBI | CHEBI:35890 |
IUPAC Name | trimethylphosphane |
SMILES | CP(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3014303
|
Thermo Scientific Chemicals
03014303 |
1 g |
Each for $92.14
|
|
|||||
AA3014306
|
Thermo Scientific Chemicals
03014306 |
5 g |
Each for $316.93
|
|
|||||
AA3014314
|
Thermo Scientific Chemicals
03014314 |
25 g |
Each for $1,285.23
|
|
|||||
Description
It is used in the preparation of organic phosphorus compounds and frequently-used phosphine ligand. It is also a highly basic ligand that forms complexes with most metals. In addition, it can react with bromomethyl-benzene to get benzyl-trimethyl-phosphonium; bromide.Â
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
594-09-2 | |
76.079 | |
YWWDBCBWQNCYNR-UHFFFAOYSA-N | |
68983 | |
trimethylphosphane |
C3H9P | |
MFCD00008510 | |
trimethylphosphine, trimethyl phosphine, pme3, trimethylphosphorus, phosphine, trimethyl, me3p, ch3 3p, trimethylphospine, trimethylphosphine?, phosphine,trimethyl | |
CHEBI:35890 | |
CP(C)C |
Specifications
594-09-2 | |
38°C to 40°C | |
Pungent | |
MFCD00008510 | |
UN2845 | |
trimethylphosphine, trimethyl phosphine, pme3, trimethylphosphorus, phosphine, trimethyl, me3p, ch3 3p, trimethylphospine, trimethylphosphine?, phosphine,trimethyl | |
YWWDBCBWQNCYNR-UHFFFAOYSA-N | |
trimethylphosphane | |
68983 | |
76.08 | |
Liquid |
0.735 g/mL | |
-30°C (-22°F) | |
C3H9P | |
1 g | |
Air and moisture sensitive | |
Soluble in water. | |
CP(C)C | |
76.079 | |
CHEBI:35890 | |
99% | |
Trimethylphosphine |
Safety and Handling
GHS H Statement
H225-H250-H252-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P222-P233-P235-P235+P410-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H250-H252-H314-H335
DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 209-823-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only