missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Trimethylsilyl)methylmagnesium chloride, 0.5M in 2-MeTHF, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H54635AE

Catalog No. AAH54635AE


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(Trimethylsilyl)methylmagnesium chloride
C4H11ClMgSi
MFCD00009916
UN2924
trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride
C[Si](C)(C)C[Mg]Cl
146.97
146.97
13170-43-9
(CH3)3SiCH2MgCl
100 mL
Air and moisture sensitive
NAQATMJWCJCHOZ-UHFFFAOYSA-M
magnesium;methanidyl(trimethyl)silane;chloride
2734900
0.5M in 2-MeTHF

Chemical Identifiers

13170-43-9
146.97
NAQATMJWCJCHOZ-UHFFFAOYSA-M
2734900
C[Si](C)(C)C[Mg]Cl
C4H11ClMgSi
MFCD00009916
trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride
magnesium;methanidyl(trimethyl)silane;chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H260-H302-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only