Learn More
(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
CAS: 13170-43-9 | C4H11ClMgSi | 146.97 g/mol
$283.16 - $283.16
Chemical Identifiers
| CAS | 13170-43-9 |
|---|---|
| Molecular Formula | C4H11ClMgSi |
| Molecular Weight (g/mol) | 146.97 |
| MDL Number | MFCD00009916 |
| InChI Key | NAQATMJWCJCHOZ-UHFFFAOYSA-M |
| Synonym | trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride |
| PubChem CID | 2734900 |
| SMILES | C[Si](C)(C)C[Mg]Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC377461000
|
Thermo Scientific Chemicals
377461000 |
100 mL | AcroSeal™ Glass bottle |
Each for $283.16
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13170-43-9 | |
| 146.97 | |
| NAQATMJWCJCHOZ-UHFFFAOYSA-M | |
| 2734900 |
| C4H11ClMgSi | |
| MFCD00009916 | |
| trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride | |
| C[Si](C)(C)C[Mg]Cl |
Specifications
| Colorless to Green | |
| 18.37,80.22 | |
| C4H11ClMgSi | |
| MFCD00009916 | |
| 16,118; 15,343; 12,540; 06,636; 05,724; 10,433 | |
| trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride | |
| NAQATMJWCJCHOZ-UHFFFAOYSA-M | |
| [(chloromagnesio)methyl]trimethylsilane | |
| 2734900 | |
| AcroSeal™ Glass bottle | |
| 100 mL | |
| Low Melting Solid |
| 13170-43-9,109-99-9,108-88-3 | |
| 19.78,81.63 | |
| (CH3)3Si-CH2-MgCl | |
| 1.2 to 1.5M | |
| 1.0400g/mL | |
| Solubility in water: reacts violently with water. | |
| C[Si](C)(C)C[Mg]Cl | |
| 146.97 | |
| 146.97 | |
| 1.04 | |
| −17°C | |
| (Trimethylsilyl)methylmagnesium chloride, 1.1M Solution in THF |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. Harmful if swallowed. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
RUO – Research Use Only