missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylsilyldiazomethane (ca. 10% in Hexane, ca. 0.6mol/L), TCI America™
Supplier: TCI America T114610ML
Specifications
Trimethylsilyldiazomethane (ca. 10% in Hexane, ca. 0.6mol/L) | |
Yellow | |
MFCD00053946 | |
2924 | |
ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
diazomethyl(trimethyl)silane | |
167693 | |
Liquid |
18107-18-1 | |
C4H10N2Si | |
10 mL | |
trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4 | |
C[Si](C)(C)C=[N+]=[N-] | |
114.223 | |
114.22 |
Chemical Identifiers
18107-18-1 | |
114.223 | |
ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
167693 | |
C[Si](C)(C)C=[N+]=[N-] |
C4H10N2Si | |
MFCD00053946 | |
trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4 | |
diazomethyl(trimethyl)silane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator