missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsulfonium bromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2509814

 View more versions of this product

Catalog No. AAB2509814

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trimethylsulfonium bromide
203°C (sublimation)
MFCD00051367
Hygroscopic
GOTIICCWNAPLMN-UHFFFAOYSA-M
trimethylsulfanium;bromide
2735154
98%
3084-53-5
C3H9BrS
25 g
trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f
C[S+](C)C.[Br-]
157.069
157.07

Chemical Identifiers

3084-53-5
157.069
GOTIICCWNAPLMN-UHFFFAOYSA-M
2735154
C[S+](C)C.[Br-]
C3H9BrS
MFCD00051367
trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f
trimethylsulfanium;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only