missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylsulfonium Iodide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140250250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylsulfonium iodide | |
215.0°C to 220.0°C | |
Authentic | |
Plastic bottle | |
(CH3)3SI | |
25 g | |
trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340 | |
VFJYIHQDILEQNR-UHFFFAOYSA-M | |
trimethylsulfanium;iodide | |
75127 | |
98% |
2181-42-2 | |
White to Yellow | |
97.5% min. (Argentometry) | |
C3H9IS | |
MFCD00011632 | |
01,314; 02,169; 01,1236; 02,171; 13,15 | |
Solubility in water: soluble | |
C[S+](C)C.[I-] | |
204.07 | |
204.07 | |
Crystalline Powder, Crystals or Chunks |
Chemical Identifiers
2181-42-2 | |
204.07 | |
VFJYIHQDILEQNR-UHFFFAOYSA-M | |
75127 | |
C[S+](C)C.[I-] |
C3H9IS | |
MFCD00011632 | |
trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340 | |
trimethylsulfanium;iodide |
Safety and Handling
EINECSNumber : 218-555-4
RUO – Research Use Only