Learn More
Trimethylsulfoxonium iodide, 98%, Thermo Scientific Chemicals
$96.24 - $96.24
Chemical Identifiers
CAS | 1774-47-6 |
---|---|
Molecular Formula | C3H9IOS |
Molecular Weight (g/mol) | 220.07 |
MDL Number | MFCD00011899 |
InChI Key | BPLKQGGAXWRFOE-UHFFFAOYSA-M |
Synonym | trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1 |
PubChem CID | 74498 |
IUPAC Name | trimethyl(oxo)-λâ¶-sulfanylium iodide |
SMILES | [I-].C[S+](C)(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC140261000
|
Thermo Scientific Chemicals
140261000 |
100 g | Plastic bottle |
Each for $96.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1774-47-6 | |
220.07 | |
BPLKQGGAXWRFOE-UHFFFAOYSA-M | |
74498 | |
[I-].C[S+](C)(C)=O |
C3H9IOS | |
MFCD00011899 | |
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1 | |
trimethyl(oxo)-λâ¶-sulfanylium iodide |
Specifications
1774-47-6 | |
White to Yellow | |
98% | |
C3H9IOS | |
MFCD00011899 | |
trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1 | |
BPLKQGGAXWRFOE-UHFFFAOYSA-M | |
trimethyl(oxo)-λâ¶-sulfanylium iodide | |
74498 | |
98% | |
Trimethylsulfoxonium iodide, 98% |
175.0°C | |
Authentic | |
Plastic bottle | |
(CH3)3S(O)I | |
100 g | |
Solubility in water: 15g/L (20°C). Other solubilities: insoluble in ethanol and ether | |
[I-].C[S+](C)(C)=O | |
220.07 | |
220.07 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 217-204-2
RTECSNumber : WS3585000
TSCA : TSCA
RUO – Research Use Only