missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethyltin Chloride 98.0+%, TCI America™
$148.13 - $454.25
Chemical Identifiers
CAS | 1066-45-1 |
---|---|
Molecular Formula | C3H11ClSn |
Molecular Weight (g/mol) | 201.28 |
MDL Number | MFCD00000520 |
InChI Key | FVFGWISLDLUGHX-UHFFFAOYSA-N |
Synonym | trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride |
PubChem CID | 14016 |
IUPAC Name | trimethyltin chloride |
SMILES | Cl.C[SnH](C)C |
Chemical Identifiers
1066-45-1 | |
201.28 | |
FVFGWISLDLUGHX-UHFFFAOYSA-N | |
14016 | |
Cl.C[SnH](C)C |
C3H11ClSn | |
MFCD00000520 | |
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride | |
trimethyltin chloride |
Specifications
1066-45-1 | |
White | |
C3H11ClSn | |
5 g | |
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride | |
Cl.C[SnH](C)C | |
201.28 | |
199.27 | |
Crystalline Powder |
40°C | |
156°C | |
MFCD00000520 | |
3146 | |
FVFGWISLDLUGHX-UHFFFAOYSA-N | |
trimethyltin chloride | |
14016 | |
≥98.0% (GC) | |
Trimethyltin Chloride |
Safety and Handling
RTECSNumber : WH6850000
TSCA : Yes
Recommended Storage : Refrigerator