missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylvinylammonium Bromide 97.0+%, TCI America™

Supplier:  TCI America T04941G

 View more versions of this product

Catalog No. T04941G


Explore available promotions

Description

Description

Specifications

Specifications

Trimethylvinylammonium Bromide
White-Yellow
MFCD00050204
Neurine Bromide
C[N+](C)(C)C=C.Br
167.07
166.06
Crystalline Powder
10603-92-6
C5H13BrN+
1 g
IDVSELVVGYIOEX-UHFFFAOYSA-N
ethenyl(trimethyl)azanium;hydrobromide
57345485
≥97.0% (T)

Chemical Identifiers

10603-92-6
167.07
IDVSELVVGYIOEX-UHFFFAOYSA-N
57345485
C[N+](C)(C)C=C.Br
C5H13BrN+
MFCD00050204
Neurine Bromide
ethenyl(trimethyl)azanium;hydrobromide
Videos
Safety and Handling

Safety and Handling

RTECSNumber : BT3675000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions