Learn More
Triolein, tech., Thermo Scientific Chemicals
$284.40 - $828.21
Chemical Identifiers
CAS | 122-32-7 |
---|---|
Molecular Formula | C57H104O6 |
Molecular Weight (g/mol) | 885.453 |
MDL Number | MFCD00137563 |
InChI Key | PHYFQTYBJUILEZ-BSCDBXJPSA-N |
Synonym | triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate |
PubChem CID | 45253964 |
IUPAC Name | [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate |
SMILES | CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6241914
|
Thermo Scientific Chemicals
J6241914 |
25 g |
Each for $284.40
|
|
|||||
AAJ6241922
|
Thermo Scientific Chemicals
J6241922 |
100 g |
Each for $828.21
|
|
|||||
Description
Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.
Solubility
Soluble in chlorofom, ether, and carbon tetrachloride
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C
Chemical Identifiers
122-32-7 | |
885.453 | |
PHYFQTYBJUILEZ-BSCDBXJPSA-N | |
45253964 | |
CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC |
C57H104O6 | |
MFCD00137563 | |
triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate | |
[2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate |
Specifications
122-32-7 | |
0.91 | |
330°C (626°F) | |
1.468 | |
MFCD00137563 | |
1718692 | |
triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate | |
PHYFQTYBJUILEZ-BSCDBXJPSA-N | |
[2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate | |
45253964 | |
75% | |
Liquid |
Yellow | |
235°C to 240°C (18 mmHg) | |
C57H104O6 | |
(C17H33COOCH2)2CHOCOC17H33 | |
25 g | |
14,9732 | |
Soluble in chlorofom, ether, and carbon tetrachloride | |
CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC | |
885.453 | |
885.45 | |
Technical | |
Triolein |
Safety and Handling
EINECSNumber : 204-534-7
RTECSNumber : RG1936500
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only