missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trioleyl Phosphite (so called) 98.0+%, TCI America™

$53.32 - $321.92

Chemical Identifiers

CAS 13023-13-7
Molecular Formula C54H105O3P
Molecular Weight (g/mol) 833.405
MDL Number MFCD00059452
InChI Key QULKPMBZSVVAJM-UHFFFAOYSA-N
Synonym Phosphorous Acid Trioleyl Ester
PubChem CID 54247678
IUPAC Name tris(octadec-9-enyl) phosphite
SMILES CCCCCCCCC=CCCCCCCCCOP(OCCCCCCCCC=CCCCCCCCC)OCCCCCCCCC=CCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P039025ML
View Documents
TCI America
P039025ML
25 mL
Each for $53.32
 
P0390500ML
View Documents
TCI America
P0390500ML
500 mL
Each for $321.92
 
Description

Description

Specifications

Chemical Identifiers

13023-13-7
833.405
QULKPMBZSVVAJM-UHFFFAOYSA-N
54247678
CCCCCCCCC=CCCCCCCCCOP(OCCCCCCCCC=CCCCCCCCC)OCCCCCCCCC=CCCCCCCCC
C54H105O3P
MFCD00059452
Phosphorous Acid Trioleyl Ester
tris(octadec-9-enyl) phosphite

Specifications

13023-13-7
C54H105O3P
25 mL
QULKPMBZSVVAJM-UHFFFAOYSA-N
tris(octadec-9-enyl) phosphite
54247678
≥98.0% (T)
Trioleyl Phosphite (so called)
Yellow
MFCD00059452
Phosphorous Acid Trioleyl Ester
CCCCCCCCC=CCCCCCCCCOP(OCCCCCCCCC=CCCCCCCCC)OCCCCCCCCC=CCCCCCCCC
833.405
833.40
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'