Learn More
Triphenyl phosphate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0813030
Description
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.
Solubility
Reacts with water.
Notes
Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
Specifications
Triphenyl phosphate | |
48°C to 52°C | |
223°C (433°F) | |
C18H15O4P | |
250 g | |
1888236 | |
triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech | |
XZZNDPSIHUTMOC-UHFFFAOYSA-N | |
triphenyl phosphate | |
8289 | |
326.29 |
115-86-6 | |
247°C to 250°C (11 mmHg) | |
Odorless | |
MFCD00003031 | |
UN3077 | |
14,9742 | |
Reacts with water. | |
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1 | |
326.29 | |
CHEBI:35033 | |
98% |
Chemical Identifiers
115-86-6 | |
326.29 | |
XZZNDPSIHUTMOC-UHFFFAOYSA-N | |
8289 | |
triphenyl phosphate |
C18H15O4P | |
MFCD00003031 | |
triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech | |
CHEBI:35033 | |
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1 |
Safety and Handling
P260-P314-P501c
H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 204-112-2
RTECSNumber : TC8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only