missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triphenylene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140360010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triphenylene | |
217-59-4 | |
White to Beige | |
Authentic | |
Glass bottle | |
MFCD00001108 | |
05, 720 | |
9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl | |
C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21 | |
228.29 | |
CHEBI:33080 | |
98% |
98% | |
196.0°C to 200.0°C | |
438.0°C | |
97.5% min. (HPLC) | |
C18H12 | |
1 g | |
15, 9913 | |
SLGBZMMZGDRARJ-UHFFFAOYSA-N | |
triphenylene | |
9170 | |
228.29 | |
Crystalline Needles |
Chemical Identifiers
217-59-4 | |
228.29 | |
SLGBZMMZGDRARJ-UHFFFAOYSA-N | |
9170 | |
triphenylene |
C18H12 | |
MFCD00001108 | |
9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl | |
CHEBI:33080 | |
C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21 |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 205-922-9
RTECSNumber : YK2925000
RUO – Research Use Only