missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethane, 99+%

CAS: 519-73-3 | C19H16 | 244.34 g/mol

$67.38 - $67.38

Chemical Identifiers

CAS 519-73-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00004763
InChI Key AAAQKTZKLRYKHR-UHFFFAOYSA-N
Synonym triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
PubChem CID 10614
ChEBI CHEBI:76212
IUPAC Name benzhydrylbenzene
SMILES C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC146380250
SDS View Documents
Thermo Scientific Chemicals
146380250
25 g Glass bottle
Each for $67.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

519-73-3
244.34
AAAQKTZKLRYKHR-UHFFFAOYSA-N
10614
benzhydrylbenzene
C19H16
MFCD00004763
triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
CHEBI:76212
C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

519-73-3
Yellow
358.0°C to 359.0°C (754.0 mmHg)
99% min. (GC)
C19H16
MFCD00004763
05,698
15,9914
Solubility in water: insoluble in water
C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1
244.34
CHEBI:76212
99+%
Triphenylmethane, 99+%
90.0°C to 94.0°C
1.0100g/mL
Authentic
Glass bottle
(C6H5)3CH
25 g
1.01
triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
AAAQKTZKLRYKHR-UHFFFAOYSA-N
benzhydrylbenzene
10614
244.34
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-275-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only