missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate, tech. 85%, Thermo Scientific Chemicals

In the purification of precious metals

$277.58 - $878.40

Chemical Identifiers

CAS 335421-90-4
Molecular Formula C18H12Na3O9PS3
Molecular Weight (g/mol) 568.41
MDL Number MFCD00145472
InChI Key MYAJTCUQMQREFZ-UHFFFAOYSA-K
Synonym sodium 3,3',3-phosphinetriyltribenzenesulfonate, tppts, triphenylphosphine-3,3',3-trisulfonic acid trisodium salt, tris 3-sulfophenyl phosphine trisodium salt, trisodium 3,3',3-phosphinetriyltris benzene-1-sulphonate, trisodium 3-bis 3-sulfonatophenyl phosphanylbenzenesulfonate, tri m-sulfophenyl phosphine, sodium salt, benzenesulfonic acid, 3,3',3-phosphinidynetris-, trisodium salt, triphenylphosphine-3,3 ,3-trisulfonic acid trisodium salt, pubchem16013
PubChem CID 6099338
IUPAC Name trisodium;3-bis(3-sulfonatophenyl)phosphanylbenzenesulfonate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3953801
View Documents
Thermo Scientific Chemicals
03953801
0.25 g
Each for $277.58
 
AA3953803
View Documents
Thermo Scientific Chemicals
03953803
1 g
Each for $878.40
 
Description

Description

Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate is useful for the preparation of water-soluble organometal catalysts, which finds application in the hydroformylation of propene. It acts as a Lewis base, which is stronger than triphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

335421-90-4
568.41
MYAJTCUQMQREFZ-UHFFFAOYSA-K
6099338
C18H12Na3O9PS3
MFCD00145472
sodium 3,3',3-phosphinetriyltribenzenesulfonate, tppts, triphenylphosphine-3,3',3-trisulfonic acid trisodium salt, tris 3-sulfophenyl phosphine trisodium salt, trisodium 3,3',3-phosphinetriyltris benzene-1-sulphonate, trisodium 3-bis 3-sulfonatophenyl phosphanylbenzenesulfonate, tri m-sulfophenyl phosphine, sodium salt, benzenesulfonic acid, 3,3',3-phosphinidynetris-, trisodium salt, triphenylphosphine-3,3 ,3-trisulfonic acid trisodium salt, pubchem16013
trisodium;3-bis(3-sulfonatophenyl)phosphanylbenzenesulfonate

Specifications

335421-90-4
MFCD00145472
3584807
Soluble in water.
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)P(C1=CC(=CC=C1)S([O-])(=O)=O)C1=CC(=CC=C1)S([O-])(=O)=O
568.41
568.41 (Anhydrous)
Technical
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate
C18H12Na3O9PS3
0.25 g
sodium 3,3',3-phosphinetriyltribenzenesulfonate, tppts, triphenylphosphine-3,3',3-trisulfonic acid trisodium salt, tris 3-sulfophenyl phosphine trisodium salt, trisodium 3,3',3-phosphinetriyltris benzene-1-sulphonate, trisodium 3-bis 3-sulfonatophenyl phosphanylbenzenesulfonate, tri m-sulfophenyl phosphine, sodium salt, benzenesulfonic acid, 3,3',3-phosphinidynetris-, trisodium salt, triphenylphosphine-3,3 ,3-trisulfonic acid trisodium salt, pubchem16013
MYAJTCUQMQREFZ-UHFFFAOYSA-K
trisodium;3-bis(3-sulfonatophenyl)phosphanylbenzenesulfonate
6099338
85%
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 264-596-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only