missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylphosphine Sulfide, 99+%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 159030100

Catalog No. AC159030100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Triphenylphosphine sulfide
3878-45-3
99+%
C18H15PS
MFCD00004917
16, 784
triphenylphosphine sulfide, e5-phosphanethione, phosphine sulfide, triphenyl, triphenylphosphine monosulfide, triphenylphosphine monosulfide, triphenylphosphino-1-thione, triphenylphosphino-1-thione, triphenylphosphorus sulfide, triphenylphosphorus sulfide, wln: spr&r&r, triphenyl-
VYNGFCUGSYEOOZ-UHFFFAOYSA-N
triphenyl(sulfanylidene)-$l^{5}-phosphane
19758
99+%
99+%
162.0°C to 164.0°C
Glass bottle
(C6H5)3P(S)
10 g
05,732
Solubility in water: practically insoluble. Other solubilities: slightly soluble in organic solvents
S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
294.35
294.34

Chemical Identifiers

3878-45-3
294.35
VYNGFCUGSYEOOZ-UHFFFAOYSA-N
19758
S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C18H15PS
MFCD00004917
triphenylphosphine sulfide, e5-phosphanethione, phosphine sulfide, triphenyl, triphenylphosphine monosulfide, triphenylphosphine monosulfide, triphenylphosphino-1-thione, triphenylphosphino-1-thione, triphenylphosphorus sulfide, triphenylphosphorus sulfide, wln: spr&r&r, triphenyl-
triphenyl(sulfanylidene)-$l^{5}-phosphane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 223-407-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only