Innovatek Triple Track Specimen Bags
Innovatek Triple Track Specimen Bags with attached outer requisition pouch
Supplier: Innovatek 4180SB9123
Description
- Unique heavy track for easy opening and positive closing
- Separate pouch outside for requisitions
- Updated to WHMIS 2015 Standards Biohazard Symbol
- Complete English and French instructions printed on bag
- Plain bags without printing also available
Specifications
No Biohazard Logo | |
Outer Requisition Pouch | |
9 x 12 in. |
Triple Track | |
500 Bags/Box |