American Educational Products Tripod Base Support Stands
Heavy-duty, tripod base support stands provide years of solid support in the laboratory.
$68.51 - $68.51
Specifications
Material Cast Iron
Product Type Tripod Base Support Stand
Finish Baked Black Enamel
For Use With (Application) Provide years of solid support in the laboratory
Quantity Each
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (English) Rod Finish Length (English) Rod Price Quantity  
Catalog Number Mfr. No. Diameter (English) Rod Finish Length (English) Rod Price Quantity  
S47809
View Documents
American Educational Products
H21300
0.3 in. Baked Black Enamel 18 in.
Each for $68.51
Description
Description
  • Available in various lengths and rod diameters
  • Feature threaded bases and threaded rods for extra stability
  • Bases constructed of rugged cast-iron and finished with chemical-resistant, baked black enamel for added stability and durability
Specifications
Specifications
Cast Iron
Baked Black Enamel
Each
Tripod Base Support Stand
Provide years of solid support in the laboratory
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'