missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tripropionin, 97%, Thermo Scientific Chemicals

$108.00 - $108.00

Chemical Identifiers

CAS 139-45-7
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00027005
InChI Key YZWRNSARCRTXDS-UHFFFAOYSA-N
Synonym tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
PubChem CID 8763
IUPAC Name 2,3-di(propanoyloxy)propyl propanoate
SMILES CCC(=O)OCC(COC(=O)CC)OC(=O)CC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC275102500
SDS View Documents
Thermo Scientific Chemicals
275102500
250 g Glass bottle
Each for $108.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

139-45-7
260.28
YZWRNSARCRTXDS-UHFFFAOYSA-N
8763
CCC(=O)OCC(COC(=O)CC)OC(=O)CC
C12H20O6
MFCD00027005
tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
2,3-di(propanoyloxy)propyl propanoate

Specifications

139-45-7
1.0820g/mL
162°C
96% min. (GC)
C12H20O6
MFCD00027005
1.082
YZWRNSARCRTXDS-UHFFFAOYSA-N
2,3-di(propanoyloxy)propyl propanoate
8763
97%
Tripropionin, 97%
Colorless
151°C (2.0 mmHg)
Authentic
Glass bottle
1.4310 to 1.434
250 g
tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
CCC(=O)OCC(COC(=O)CC)OC(=O)CC
260.28
260.28
Liquid
Videos
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 205-365-1

RTECSNumber : YK6000000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only