missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tripropylamine, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 162690025

Catalog No. AC162690025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tripropylamine
0.7600g/mL
32.5°C
0.2% max. dipropylamine (GC)
C9H21N
(CH3CH2CH2)3N
2.5L
0.76
Solubility in water: 2.6g/L (20°C). Other solubilities: soluble in most organic solvents
CCCN(CCC)CCC
143.27
0.627 mPa.s (25°C)
143.27
102-69-2
156.0°C
Authentic
Glass bottle
1.4155 to 1.4175
MFCD00009363
04, II, 623
tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
YFTHZRPMJXBUME-UHFFFAOYSA-N
N,N-dipropylpropan-1-amine
7616
CHEBI:38880
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
May cause respiratory irritation.
Harmful to aquatic life wi

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several min

GHS Signal Word: Danger

EINECSNumber : 203-047-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only