missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tripropylene Glycol Monomethyl Ether (mixture of isomer) 93.0+%, TCI America™
$29.86 - $61.38
Chemical Identifiers
CAS | 25498-49-1 |
---|---|
Molecular Formula | C10H22O4 |
MDL Number | MFCD00059142 |
Chemical Identifiers
25498-49-1 | |
C10H22O4 | |
MFCD00059142 |
Specifications
25498-49-1 | |
Yellow | |
C10H22O4 | |
25 g | |
≥93.0% (GC) | |
Tripropylene Glycol Monomethyl Ether (mixture of isomer) |
-79°C | |
243°C | |
MFCD00059142 | |
206.28 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0438&(7)-0097
RTECSNumber : UB8070000
TSCA : Yes