missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals

$107.29 - $107.29

Chemical Identifiers

CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
Synonym ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
PubChem CID 84567
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC422251000
SDS View Documents
Thermo Scientific Chemicals
422251000
100 mL Glass bottle
Each for $107.29
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

14634-91-4
692.53
CIWXFRVOSDNDJZ-UHFFFAOYSA-L
84567
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
C36H24FeN6O4S
MFCD00036428
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
iron(2+);1,10-phenanthroline;sulfate

Specifications

14634-91-4
1.0060g/mL
C36H24FeN6O4S
100 mL
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
692.53
692.53
Tris(1, 10-phenanthroline)iron(II) sulfate, 0.025M Aqueous Solution
Red
Glass bottle
MFCD00036428
1.006
CIWXFRVOSDNDJZ-UHFFFAOYSA-L
iron(2+);1,10-phenanthroline;sulfate
84567
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 238-676-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only