missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(1,10-phenanthroline)ruthenium(II) Bis(hexafluorophosphate) 98.0+%, TCI America™
Supplier: TCI America T3208200MG
Specifications
Tris(1,10-phenanthroline)ruthenium(II) Bis(hexafluorophosphate) | |
C36H24F12N6P2Ru | |
YRYUXGTVQZIGNQ-UHFFFAOYSA-N | |
1,10-phenanthroline;ruthenium(2+);dihexafluorophosphate | |
15198714 | |
≥98.0% (HPLC) |
60804-75-3 | |
200 mg | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.[Ru+2] | |
931.628 | |
931.63 | |
Crystalline Powder |
Chemical Identifiers
60804-75-3 | |
931.628 | |
15198714 | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.[Ru+2] |
C36H24F12N6P2Ru | |
YRYUXGTVQZIGNQ-UHFFFAOYSA-N | |
1,10-phenanthroline;ruthenium(2+);dihexafluorophosphate |
Safety and Handling
TSCA : No