missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(1-pyrazolyl)methane 98.0+%, TCI America™
Supplier: TCI America T30261G
Specifications
Tris(1-pyrazolyl)methane | |
104°C | |
C10H10N6 | |
1 g | |
C1=CN(N=C1)C(N1C=CC=N1)N1C=CC=N1 | |
214.23 | |
214.23 | |
Crystalline Powder |
80510-03-8 | |
White-Yellow | |
MFCD00040255 | |
QOHVIMQQEFHOLE-UHFFFAOYSA-N | |
1-[bis(1H-pyrazol-1-yl)methyl]-1H-pyrazole | |
5243808 | |
≥98.0% (GC) |
Chemical Identifiers
80510-03-8 | |
214.23 | |
QOHVIMQQEFHOLE-UHFFFAOYSA-N | |
1-[bis(1H-pyrazol-1-yl)methyl]-1H-pyrazole |
C10H10N6 | |
MFCD00040255 | |
5243808 | |
C1=CN(N=C1)C(N1C=CC=N1)N1C=CC=N1 |
Safety and Handling
TSCA : No