missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gadolinium(III), Thermo Scientific Chemicals

$111.58 - $423.67

Identifiants chimiques

CAS 14768-15-1
Molecular Formula C33H57GdO6
Molecular Weight (g/mol) 707.06
MDL Number MFCD00010349
InChI Key UIVVGOMXFUOLAN-UHFFFAOYSA-N
Synonym tris 2,2,6,6-tetramethyl-3,5-heptanedionate gadolinium
PubChem CID 132984032
IUPAC Name gadolinium;(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
SMILES [Gd+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA1448603
Afficher les documents
Thermo Scientific Chemicals
01448603
1 g
chaque for $111.58
 
AA1448606
Afficher les documents
Thermo Scientific Chemicals
01448606
5 g
chaque for $423.67
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

14768-15-1
707.06
UIVVGOMXFUOLAN-UHFFFAOYSA-N
132984032
[Gd+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
C33H57GdO6
MFCD00010349
tris 2,2,6,6-tetramethyl-3,5-heptanedionate gadolinium
gadolinium;(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one

Spécifications

14768-15-1
C33H57GdO6
1 g
tris 2,2,6,6-tetramethyl-3,5-heptanedionate gadolinium
[Gd+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
707.06
707.07
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gadolinium(III)
167°C to 170°C
MFCD00010349
Hygroscopic
UIVVGOMXFUOLAN-UHFFFAOYSA-N
gadolinium;(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
132984032
Crystalline
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 238-834-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only