missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(2-butoxyethyl) phosphate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 157240050

 View more versions of this product

Catalog No. AC157240050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tris(2-butoxyethyl) phosphate
1.0000g/mL
132°C
94% min. (GC)
C18H39O7P
[CH3(CH2)3OCH2CH2O]3P(O)
5g
tris 2-butoxyethyl phosphate, tbep, phosflex t-bep, tris butoxyethyl phosphate, tributoxyethyl phosphate, tri butoxyethyl phosphate, kronitex kp-140, tributyl cellosolve phosphate, ethanol, 2-butoxy-, phosphate 3:1, tri 2-butoxyethyl phosphate
WTLBZVNBAKMVDP-UHFFFAOYSA-N
tris(2-butoxyethyl) phosphate
6540
398.47
78-51-3
215°C to 228°C (4.0mmHg)
Authentic
Glass bottle
1.4370 to 1.439
MFCD00009456
1
Solubility in water: 1.3g/L (20°C)
CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC
398.48
CHEBI:35038
95%

Chemical Identifiers

78-51-3
398.48
WTLBZVNBAKMVDP-UHFFFAOYSA-N
6540
tris(2-butoxyethyl) phosphate
C18H39O7P
MFCD00009456
tris 2-butoxyethyl phosphate, tbep, phosflex t-bep, tris butoxyethyl phosphate, tributoxyethyl phosphate, tri butoxyethyl phosphate, kronitex kp-140, tributyl cellosolve phosphate, ethanol, 2-butoxy-, phosphate 3:1, tri 2-butoxyethyl phosphate
CHEBI:35038
CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-122-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only