Molecular Probes™ Tris-(2-Carboxyethyl)phosphine, Hydrochloride (TCEP)
Tris-(2-Carboxyethyl)phosphine, Hydrochloride (TCEP)
Supplier:  Molecular Probes™ T2556
Catalog No. T2556
Explore available promotions
Description
Description
Disufide crosslinks of cystines in proteins can be reduced to cysteine residues by TCEP (tris-(2-carboxyethyl)phosphine). Unlike DTT (dithiothreitol), TCEP does not contain thiols and therefore usually does not need to be removed prior to thiol modification.
Specifications
Specifications
Store at room temperature.
Room Temperature
Solid
1 g
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.