missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-pyridylmethyl)amine 98.0+%, TCI America™
$303.84 - $961.86
Chemical Identifiers
| CAS | 16858-01-8 |
|---|---|
| Molecular Formula | C18H18N4 |
| Molecular Weight (g/mol) | 290.37 |
| MDL Number | MFCD14708176 |
| InChI Key | VGUWFGWZSVLROP-UHFFFAOYSA-N |
| PubChem CID | 379259 |
| IUPAC Name | tris[(pyridin-2-yl)methyl]amine |
| SMILES | C(N(CC1=CC=CC=N1)CC1=CC=CC=N1)C1=CC=CC=N1 |
Chemical Identifiers
| 16858-01-8 | |
| 290.37 | |
| VGUWFGWZSVLROP-UHFFFAOYSA-N | |
| tris[(pyridin-2-yl)methyl]amine |
| C18H18N4 | |
| MFCD14708176 | |
| 379259 | |
| C(N(CC1=CC=CC=N1)CC1=CC=CC=N1)C1=CC=CC=N1 |
Specifications
| 16858-01-8 | |
| White-Yellow | |
| MFCD14708176 | |
| VGUWFGWZSVLROP-UHFFFAOYSA-N | |
| tris[(pyridin-2-yl)methyl]amine | |
| 379259 | |
| ≥98.0% (HPLC) | |
| Tris(2-pyridylmethyl)amine |
| 86°C | |
| C18H18N4 | |
| 1 g | |
| C(N(CC1=CC=CC=N1)CC1=CC=CC=N1)C1=CC=CC=N1 | |
| 290.37 | |
| 290.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : No