Learn More
Tris(4-bromophenyl)amine, 98%, Thermo Scientific Chemicals
$229.49 - $229.49
Chemical Identifiers
| CAS | 4316-58-9 |
|---|---|
| Molecular Formula | C18H12Br3N |
| Molecular Weight (g/mol) | 482.013 |
| MDL Number | MFCD00009665 |
| InChI Key | ZRXVCYGHAUGABY-UHFFFAOYSA-N |
| Synonym | tris 4-bromophenyl amine, tris p-bromophenyl amine, tris-4-bromophenyl amine, 4-bromo-n,n-bis 4-bromophenyl aniline, benzenamine, 4-bromo-n,n-bis 4-bromophenyl, 4,4',4-tribromotriphenylamine, triphenylamine, 4,4',4-tribromo, pubchem13773, tri 4-bromophenyl amine, acmc-1arf9 |
| PubChem CID | 258027 |
| IUPAC Name | 4-bromo-N,N-bis(4-bromophenyl)aniline |
| SMILES | C1=CC(=CC=C1N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6086009
|
Thermo Scientific Chemicals
H6086009 |
10 g |
Each for $229.49
|
|
|||||
Description
Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Chemical Identifiers
| 4316-58-9 | |
| 482.013 | |
| ZRXVCYGHAUGABY-UHFFFAOYSA-N | |
| 258027 | |
| C1=CC(=CC=C1N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)Br |
| C18H12Br3N | |
| MFCD00009665 | |
| tris 4-bromophenyl amine, tris p-bromophenyl amine, tris-4-bromophenyl amine, 4-bromo-n,n-bis 4-bromophenyl aniline, benzenamine, 4-bromo-n,n-bis 4-bromophenyl, 4,4',4-tribromotriphenylamine, triphenylamine, 4,4',4-tribromo, pubchem13773, tri 4-bromophenyl amine, acmc-1arf9 | |
| 4-bromo-N,N-bis(4-bromophenyl)aniline |
Specifications
| 4316-58-9 | |
| C18H12Br3N | |
| 10 g | |
| Soluble in toluene. | |
| C1=CC(=CC=C1N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)Br | |
| 482.013 | |
| 482.01 | |
| Tris(4-bromophenyl)amine |
| 141°C to 143°C | |
| MFCD00009665 | |
| tris 4-bromophenyl amine, tris p-bromophenyl amine, tris-4-bromophenyl amine, 4-bromo-n,n-bis 4-bromophenyl aniline, benzenamine, 4-bromo-n,n-bis 4-bromophenyl, 4,4',4-tribromotriphenylamine, triphenylamine, 4,4',4-tribromo, pubchem13773, tri 4-bromophenyl amine, acmc-1arf9 | |
| ZRXVCYGHAUGABY-UHFFFAOYSA-N | |
| 4-bromo-N,N-bis(4-bromophenyl)aniline | |
| 258027 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only