missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris, 99 to 99.5%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215217601

 View more versions of this product

Catalog No. ICN15217601


Explore available promotions

Description

Description

Specifications

Specifications

Tris
White
C4H11NO3
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Crystalline Powder
77-86-1
99 to 99.5%
1 kg
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14

Chemical Identifiers

77-86-1
121.136
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
Videos
SDS
Documents

Documents

Promotions

Promotions